Order Entry
United States
ContactUsLinkComponent
SV Calibration Mix #7 (3 Components), Restek
SV Calibration Mix #7 (3 Components), Restek
Catalog # RK31013
Supplier:  Restek
CAS Number:  
undefined
SV Calibration Mix #7 (3 Components), Restek
Catalog # RK31013
Supplier:  Restek
Supplier Number:  31013
CAS Number:  

Specifications

  • Concentration:
    2000 µg/ml each in methylene chloride
  • Description:
    SV Calibration Mix #7
  • Size:
    1 ml/Ampule
  • Cat. No.:
    RK31013
  • Certification:
    CRM
  • Supplier no.:
    31013

Specifications

About this item

SV Calibration mix contains 1,2-dichlorobenzene, 1,3-dichlorobenzene, 1,4-dichlorobenzene.

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.