Trichlorofluoromethane (CFC-11), Restek
Catalog # RK30421
Supplier: Restek
CAS Number:
Specifications
- Concentration:2000 µg/ml in P and T methanol
- CAS No.:75-69-4
- Description:Trichlorofluoromethane (CFC-11)
- Size:1 ml/Ampule
- Formula:CFCl₃
- Description:Trichlorofluoromethane (CFC-11)
- Cat. No.:RK30421
- Certification:CRM
- Supplier no.:30421
Specifications
About this item
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.