Order Entry
United States
ContactUsLinkComponent
Methacrylonitrile, Restek
Methacrylonitrile, Restek
Catalog # RK30297
Supplier:  Restek
CAS Number:  
undefined
Methacrylonitrile, Restek
Catalog # RK30297
Supplier:  Restek
Supplier Number:  30297
CAS Number:  

Specifications

  • Concentration:
    2000 µg/ml in P and T methanol
  • CAS No.:
    126-98-7
  • Description:
    Methacrylonitrile
  • Size:
    1 ml/Ampule
  • Formula:
    C₄H₅N
  • Description:
    Methacrylonitrile
  • Cat. No.:
    RK30297
  • Certification:
    CRM
  • Supplier no.:
    30297

Specifications

About this item

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.