Order Entry
United States
ContactUsLinkComponent
Dichlorodifluoromethane (CFC-12), Restek
Dichlorodifluoromethane (CFC-12), Restek
Catalog # RK30275
Supplier:  Restek
CAS Number:  
undefined
Dichlorodifluoromethane (CFC-12), Restek
Catalog # RK30275
Supplier:  Restek
Supplier Number:  30275
CAS Number:  

Specifications

  • Concentration:
    2000 µg/ml in P and T methanol
  • CAS No.:
    75-71-8
  • Description:
    Dichlorodifluoromethane (CFC-12)
  • Size:
    1 ml/Ampule
  • Formula:
    CCl₂F₂
  • Description:
    Dichlorodifluoromethane (CFC-12)
  • Cat. No.:
    RK30275
  • Certification:
    CRM
  • Supplier no.:
    30275

Specifications

About this item

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.