504.1 Calibration Mix (3 components), Restek
Catalog # RK30239
Supplier: Restek
CAS Number:
Specifications
- Concentration:200 µg/ml each in P and T methanol
- Description:504.1 Calibration Mix
- Size:1 ml/Ampule
- Cat. No.:RK30239
- Certification:CRM
- Supplier no.:30239
Specifications
About this item
Mix contains: Chlorobenzene-d5 (3114-55-4); 1,4-Dichlorobenzene-d4 (3855-82-1); Fluorobenzene (462-06-6).
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.