PVOC Mix (California) (7 Components), Restek
Catalog # RK30231
Supplier: Restek
CAS Number:
Specifications
- Concentration:1000 µg/ml each in P and T methanol
- Description:PVOC Mix (California)
- Size:1 ml/Ampule
- Cat. No.:RK30231
- Certification:CRM
- Supplier no.:30231
Specifications
About this item
Mix contains: Benzene (71-43-2); Ethylbenzene (100-41-4); Methyl tert-butyl ether (MTBE) (1634-04-4); Toluene (108-88-3); m-Xylene (108-38-3); o-Xylene (95-47-6); p-Xylene (106-42-3).
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.