Order Entry
United States
ContactUsLinkComponent
524 Calibration Mix #7 Kit, Restek
524 Calibration Mix #7 Kit, Restek
Catalog # RK30202
Supplier:  Restek
CAS Number:  
undefined
524 Calibration Mix #7 Kit, Restek
Catalog # RK30202
Supplier:  Restek
Supplier Number:  30202
CAS Number:  

Specifications

  • Concentration:
    Contains 1 ml each of these mixtures
  • Description:
    524 Calibration Mix #7 Kit
  • Size:
    Kit
  • Cat. No.:
    RK30202
  • Certification:
    CRM
  • Supplier no.:
    30202

Specifications

About this item

Contains 1 ml each of these mixtures: 524 calibration mix #7A, 524 calibration mix #7B. calibration mix #7A contains Acetone (67-64-1), 2-Butanone (MEK) (78-93-3), 2-Hexanone (591-78-6), 4-Methyl-2-pentanone (MIBK) (108-10-1), Tetrahydrofuran (109-99-9); calibration mix #7B contains Acrylonitrile (107-13-1)
Allyl chloride (3-chloropropene) (107-05-1), Ethyl methacrylate (97-63-2), Methyl acrylate (96-33-3), Methyl methacrylate (80-62-6), Nitrobenzene (98-95-3), Pentachloroethane (76-01-7).

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.