Silylation Derivatization Reagents, Restek
: Restek
Contains: 35600, 35601: MSTFA (N-methyl-N-trimethylsilytrifluoroacetamide) (24589-78-4); 35602, 35603: MSTFA w/1% TMCS (N-methyl-N-trimethylsilytrifluoroacetamide w/1% trimethylchlorosilane) (24589-78-4); 35604, 35605: BSTFA (N,O-bis[trimethylsilyl]trifluoroacetamide) (25561-30-2); 35606, 35607: BSTFA w/1% TMCS (N,O-bis[trimethylsilyltrifluoroacetamide] w/1% trimethylchlorosilane) (25561-30-2); 35608, 35610: MTBSTFA w/1% TBDMCS (N-methyl-N[tert-butyldimethylsilyl trifluoroacetamide] w/1% tert-butyldimethylchlorosilane) (77377-52-7); 35611, 35612: TMCS (trimethylchlorosilane) (75-77-4)
- Replace active hydrogen, reducing polarity and making the compounds more volatile.
- Increase stability of derivatives.
- CAS No.:24589-78-4
- Formula:C₆H₁₂F₃NOSi
- Description:MSTFA w/1% TMCS (N-methyl-N-trimethylsilytrifluoroacetamide w/1% trimethylchlorosilane)
Frequently Bought Together