Aldehyde-Ketone-DNPH TO-11A Calibration Mix (15 components), Restek
10029-522
: Restek
:
- Concentration:15 µg/ml each in acetonitrile
- Description:Aldehyde-Ketone-DNPH TO-11A Calibration Mix
- Size:1 ml/Ampule
- Cat. No.:10029-522
- Certification:CRM
- Supplier no.:31808
This consists of acetaldehyde-DNPH; acetone-DNPH; acrolein-DNPH; benzaldehyde-DNPH; n-butyraldehyde-DNPH; crotonaldehyde-DNPH; 2,5-dimethylbenzaldehyde-DNPH; formaldehyde-DNPH; hexaldehyde-DNPH; isovaleraldehyde-DNPH; propionaldehyde-DNPH; m-tolualdehyde-DNPH; o-Tolualdehyde-DNPH; p-tolualdehyde-DNPH and aleraldehyde-DNPH.
: *The reported concentrations reflect the amount of aldehyde or ketone in the mixture. The concentration of derivatized aldehyde or ketone is not reported.