ApopTag® Plus Peroxidase Apoptosis Assay Kit, MP Biomedicals
Catalog # IC11S7101KIT
Supplier: Avantor
CAS Number:
Specifications
- Storage Conditions:Store the kit at -15 °C to -25 °C until the first use. After the first use, if the kit will be used within three months, store the TdT Enzyme at -15 °C to -25 °C and store the remaining components at 2 °C to 8 °C.
- Application:Cell Viability Analysis; Cellular Imaging; Cell Structure Analysis
- Includes:Equilibration Buffer (S7105), Reaction Buffer (S7106), TdT Enzyme (S7107), Stop/Wash Buffer (S7108), Plastic Coverslips and Anti-digoxigenin-peroxidase, ApopTag™ positive control slides, DAB Substrate, DAB Dilution Buffer, affinity purified sheep polyclonal antibody.