Order Entry
United States
ContactUsLinkComponent
CSM-Arg Powder, MP Biomedicals
CSM-Arg Powder, MP Biomedicals
Supplier:  MP Biomedicals
Certificates
undefined
CSM-Arg Powder, MP Biomedicals
Supplier:  MP Biomedicals

Specifications

  • Storage Conditions:
    Store CSM-Arg powder at ambient temperature (15-30°C). CSM powder is hygroscopic, therefore minimize exposure to air to prolong storage of powder mixture. Store sterile agar medium at 4°C. And liquid medium at 15-30°C in subdued light.
  • Sterility:
    Sterile. Autoclave at 121 °C for 25 minutes.
  • Application:
    Growth of Saccharomyces cerevisiae; Yeast Cell Culture; Gene Expression; Transformation; Gene Function; Gene Regulation; Selection of yeast transformants; For confirmation of auxotrophic phenotypes
  • pH:
    5 - 5.8(Lit.); solution: 5.5

Specifications

Frequently Bought Together