Methylene Chloride (Dichloromethane), Restek
Catalog # RK30401
Supplier: Restek
CAS Number:
Specifications
- Concentration:2000 µg/ml in P and T methanol
- CAS No.:75-09-2
- Description:Methylene chloride (dichloromethane)
- Size:1 ml/Ampule
- Formula:CH₂Cl₂
- Description:Methylene chloride (dichloromethane)
- Cat. No.:RK30401
- Certification:CRM
- Supplier no.:30401
Specifications
About this item
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.