Rudiger® Skeleton Hardware Replacement Kits
Supplier: RUDIGER ANATOMIE GMBH
Certificates
About this item
Sets of those hard to find parts.
- Replace the hardware on older skeletons
- Matches your existing hardware from Rudiger
Each of these convenient sets allows you to replace a section of hardware for your Rudiger Skeletons. Each kit contains the material to replace all the hardware at one joint.
Arm Kit: Replaces the assembly at one shoulder and contains 1 U-Bracket, 2 Thumbscrews, 1 Threaded Pin, 2 washers and 1 Screw.
Leg Kit: Replaces the assembly at the pelvis to attach 1 leg and contains 1 U-Bracket, 4 Washers, 1 Threaded Pin, 3 Thumbscrews, and 1 nut.
Knee/Elbow Kit: Includes the hardware to reattach at a knee joint and contains 1 eye screw and 1 thumbscrew.
Frequently Bought Together