

Acetone ≥99.9% (by GC, corrected for water content) for pesticide residue analysis, for spectrophotometry, Burdick & Jackson™
Catalog # BJCS011-200
Supplier: Honeywell Research Chemicals
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Size:200 L
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental Certification:Chem21 solvent selection guide recommended after discussion
- Cat. No.:BJCS011-200
- Supplier no.:CS011-200
Specifications
Specification Test Results
- Electron Capture GC:10 ng/L max.
- Water by Karl Fischer Titration:0.5 % max.
- GC Analysis (excluding water):99.9 % min.
- Refractive Index (20ºC):1.3583 - 1.3589
- Residue:1 mg/L max.
- UV Absorbance @ 340 nm:0.06 AU max.
- UV Absorbance @ 350 nm:0.01 AU max.
- UV Absorbance @ 375 nm:0.005 AU max.
- UV Absorbance @ 400 nm:0.005 AU max.
- UV Cutoff:330 nm max.
Specification Test Results
About this item
:
:58.08 g/mol
Boiling Pt:56 °C
Melting Pt:–94.8 °C
Density:0.79 g/cm3
UN:1090
ADR:3,II
:GHS02
:GHS07
CAS Number:67-64-1
Formula:CH₃COCH₃
MDL Number:MFCD00008765