40000 Count Dual Display Handheld LCR Meters
470227-630
- Description:40,000 Count Dual Display Handheld LCR Meters
- Cat. No.:470227-630
The handheld LCR meters measure inductance, capacitance, and resistance quickly and precisely.
- 40,000 counts resolution on primary and 10,000 counts resolution on secondary display
- Fast auto range design for rapid, easy component measurements
- Data Hold and min/max/average recording
- USB (Virtual COM) interface
Fast auto ranging and quick measurement configuration such as measurement parameter and test frequency selection make these meters very simple to operate. The meter also includes handy functions such as data hold, Min/Max/Average recording, tolerance sorting, and relative mode.
Measurement data can continuously transfer to a PC via the meter's mini USB interface, using either the provided data logging software or SCPI commands sent from a custom program.
Applications
Passive component troubleshooting
Electronic assembly
Quality control (component sorting)