To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Cat. No.:10763-240
- Storage temperature:+2…+8 °C
- Supplier no.:640914
- No. of tests:100
- Species reactivity:All mammalian species
Specifications
About this item
FITC Annexin V Apoptosis Detection Kit with PI is used for the identification of apoptotic and necrotic cells and is suitable for flow cytometric analysis of antibody surface-stained cells.
Annexin V (or Annexin A5) is a member of the annexin family of intracellular proteins that binds to phosphatidylserine (PS) in a calcium dependent manner. Fluorochrome-labeled Annexin V can be used to specifically target and identify apoptotic cells.