Extra Bright Laser Pointer
Extra bright laser pointer, 635 nm wavelength, 5 mW (Class IIIA U.S. CDRH)
Catalog # 470111-384
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
LASER POINTER RED CLASS 3 Catalog #470001-980
Product 470001-980 is similar to 470111-384 but manufacturer and specifications may vary.
Product Details & Documents
High-brightness, red light laser pointer with integral adjustable-focusing lens.
- Small size
- High-quality collimated laser beam
- Adjustable focus laser beam
- Beam diameter mm 1.10
- Beam divergence mRad 1.20
- Battery powered for convenient and portable operation
It produces a very bright beam of light and its 635 nm emission is a near match to the long-used helium-neon 632.8 nm emission laser. Featuring an adjustable lens, the laser beam can be focused to a fine spot for detail or "opened" to show a broader view for any type of optical experiment or presentation. This laser is a great product for educational use, audio/visual presentations.
Ordering information: Includes carrying case and two AAA batteries.
Specifications
- Description:Extra bright laser pointer, 635 nm wavelength, 5 mW (Class IIIA U.S. CDRH)