Order Entry
United States
ContactUsLinkComponent
Density: Lava Lamp
Density: Lava Lamp
Catalog # 470228-176
Supplier:  Ward's Science
undefined
Density: Lava Lamp
Catalog # 470228-176
Supplier:  Ward's Science
Supplier Number:  IS1008

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Density: Lava Lamp
  • Cat. No.:
    470228-176
  • No. of uses:
    1

Specifications

About this item

What is Density?

  • Aligned to Next Generation science standards (NGSS)
  • Developed for grades 2-5

Density is a concept that can be difficult to explain to your elementary and middle school students. Help them visualize and understand density and introduce the idea of polarity as well with this demonstration for the whole classroom.

Ordering information: Includes 5 Plastic Bottles, 1L Vegetable Oil, 25 mL Food Coloring, 10 Alka Seltzer Tablets.
Caution: WARNING CHOKING HAZARD SMALL PART. Not for children under 3 yrs.