Order Entry
United States
ContactUsLinkComponent
Formic acid ∼98% (by titrimetric analysis) for mass spectrometry, Fluka™
Formic acid ∼98% (by titrimetric analysis) for mass spectrometry, Fluka™
Catalog # BJ94318-250ML
CAS Number:  
Formic acid ∼98% (by titrimetric analysis) for mass spectrometry, Fluka™
Catalog # BJ94318-250ML
Supplier Number:  94318-250ML
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pack type:
    Glass bottle
  • Size:
    250 ml
  • Environmentally Preferable:
  • Cat. No.:
    BJ94318-250ML
  • Supplier no.:
    94318-250ML

Specifications

Specification Test Results

  • Assay:
    Min. 97.5 %
  • Suitability for the LC-MS:
    complying
  • Water (Karl Fischer):
    Max. 2.5 %
  • Non-volatile matter:
    Max. 20 ppm
  • APHA:
    Max. 15
  • Silver (Ag):
    Max. 0.02 ppm
  • Aluminium (Al):
    Max. 0.05 ppm
  • Barium (Ba):
    Max. 0.05 ppm
  • Bismuth (Bi):
    Max. 0.1 ppm
  • Calcium (Ca):
    Max. 0.2 ppm
  • Cadmium (Cd):
    Max. 0.05 ppm
  • Cobalt (Co):
    Max. 0.02 ppm
  • Chromium (Cr):
    Max. 0.05 ppm
  • Copper (Cu):
    Max. 0.02 ppm
  • Iron (Fe):
    Max. 0.2 ppm
  • Potassium (K):
    Max. 0.1 ppm
  • Lithium (Li):
    Max. 0.02 ppm
  • Magnesium (Mg):
    Max. 0.5 ppm
  • Manganese (Mn):
    Max. 0.05 ppm
  • Molybdenum (Mo):
    Max. 0.02 ppm
  • Sodium (Na):
    Max. 0.5 ppm
  • Nickel (Ni):
    Max. 0.05 ppm
  • Lead (Pb):
    Max. 0.02 ppm
  • Strontium (Sr):
    Max. 0.02 ppm
  • Thallium (Tl):
    Max. 0.05 ppm
  • Zinc (Zn):
    Max. 0.05 ppm
  • Chloride (Cl):
    Max. 5 ppm
  • Sulfate (SO₄):
    Max. 5 ppm
  • Sulfite (SO₃):
    Max. 10 ppm
  • Ammonium (NH₄):
    Max. 10 ppm
  • Appearance of the solution:
    complying

Specification Test Results

About this item

Synonyms: Aminic Acid, Methanoic acid
 :
 :46.03
Flash Pt:49.5 °C
EINECS:200-579-1
UN:1779
 :GHS05
 :GHS06
 :GHS02
Storage Temperature:Ambient
CAS Number:64-18-6
Formula:HCO₂H
MDL Number:MFCD00003297
Boiling Pt:100.4 °C (1.013 hPa)
Melting Pt:4 °C
Density:1.22 g/cm³ (20 °C)
ADR:8 II