Kimtech™ A5 Sterile Integrated Hood and Mask, Kimberly-Clark Professional
75879-282
- Clothing Size:XL
- Sterile:+
- Garment application:Cleanroom
- Color:White
- Cat. No.:75879-282
- Supplier no.:47677
This cleanroom sterile hood features a mask integrated into the garment to close gaps in protection.
- Oval head design to accommodate different hairstyles and head shapes
- Optimized viewing to limit skin exposure around forehead and cheeks
- Integrated hood and mask to close gaps in protection
The Kimtech™ sterile A5 integrated hood and mask combines two donning steps (hood and mask) into one, simplifying the donning process and further reducing the risk of contamination. This sterile product features a stretch-fit elastic hood and opening, tunneled overseams to prevent particle shedding, and Clean-Don™ technology ties for a more secure fit. An innovative design makes it easier to accommodate today's modern and diverse hairstyles comfortably and consistently.