Bacteria Lysis Kit, IBI Scientific
About this item
Supplemental kit for IBI Tri-Isolate RNA Pure Kit when working with bacterial cells.
- Tri-Isolate supplemental kit
- For extracting RNA from bacteria samples
- Contains Lysozyme and Bacterial Lysis Buffer
The Bacteria Lysis Kit is used with the IBI Tri-Isolate RNA Pure Kit when extracting RNA from bacteria samples. Bacterial cell walls are initially lysed by lysozyme, followed by the bacterial lysis buffer. The sample is then homogenized with the IBI Isolate reagent contained in the Tri-Isolate kit.
Ordering information: Bacteria Lysis Kits may be supplemented for IB47631 (100 reactions) or IB47632 (200 reactions). Applicable for bacterial cell samples only. For research and development purposes only.
2 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Supplemental kit for IBI Tri-Isolate RNA Pure Kit when working with bacterial cells.
- Tri-Isolate supplemental kit
- For extracting RNA from bacteria samples
- Contains Lysozyme and Bacterial Lysis Buffer
The Bacteria Lysis Kit is used with the IBI Tri-Isolate RNA Pure Kit when extracting RNA from bacteria samples. Bacterial cell walls are initially lysed by lysozyme, followed by the bacterial lysis buffer. The sample is then homogenized with the IBI Isolate reagent contained in the Tri-Isolate kit.
Ordering information: Bacteria Lysis Kits may be supplemented for IB47631 (100 reactions) or IB47632 (200 reactions). Applicable for bacterial cell samples only. For research and development purposes only.
Documents
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 76006-528
- 76006-530
- Description:
- Bacteria Lysis Kit
- Bacteria Lysis Kit
- Target molecule:
- DNA
- DNA
- Refine target molecule:
- Bacterial RNA
- Bacterial RNA
- Sample type:
- Bacteria
- Bacteria
- Method / Format:
- Enzymatic extraction
- Enzymatic extraction
- Prep size:
- Scalable
- Scalable
- Sample Size:
- 1 x 109 (bacteria cells)
- 1 x 109 (bacteria cells)
Specifications
Product Family Options
Product Information
- SizeAvailabilityPrice
- 76006-528Bacteria Lysis KitSupplier Number: IB47633Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.76006-528DescriptionBacteria Lysis KitTarget moleculeDNARefine target moleculeBacterial RNASample typeBacteriaMethod / FormatEnzymatic extractionPrep sizeScalableSample Size1 x 109 (bacteria cells) - 76006-530Bacteria Lysis KitSupplier Number: IB47634Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.76006-530DescriptionBacteria Lysis KitTarget moleculeDNARefine target moleculeBacterial RNASample typeBacteriaMethod / FormatEnzymatic extractionPrep sizeScalableSample Size1 x 109 (bacteria cells)



