Vibration Generator
Vibration Generator
Catalog # 470225-334
Supplier: American Scientific
Supplier #: 8420-00
Jump to:
Product Details & Documents
Mechanical representation of a sine wave signal.
- The wave drivers allows you to perform experiments with both transverse and longitudinal waves
This model can handle up to 1 Amp and has a unique locking mechanism, which keep the vibrating post secure when it is being stored. The unit can handle signals up to 10 kHz. Couple it with a digital wave generator to produce precise frequencies.
Specifications
- Description:Vibration Generator