Sine Wave Signal Generator
Catalog # 470225-332
Supplier: American Scientific
Specifications
- Description:Sine Wave Signal Generator
- Environmentally Preferable:
- Cat. No.:470225-332
- Supplier no.:8410-00
Specifications
About this item
Create waves and signals quickly and easily.
- This sine wave generator is a great economical tool for generating waves with speakers or wave drivers
It allows both the frequency (1-4,000 Hz) and amplitude of the sine wave output to be varied. The digital display features a 0.1 Hz resolution.