Order Entry
United States
ContactUsLinkComponent
Single-Stranded DNA Binding Protein (SSB) (from E. coli strain containing an overproducing clone of E. coli SSB protein.)
Single-Stranded DNA Binding Protein (SSB) (from E. coli strain containing an overproducing clone of E. coli SSB protein.)
Catalog # 75838-160
Supplier:  Avantor
CAS Number:  
undefined
Single-Stranded DNA Binding Protein (SSB) (from E. coli strain containing an overproducing clone of E. coli SSB protein.)
Catalog # 75838-160
Supplier:  Avantor
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Source:
    E. coli strain containing an overproducing clone of E. coli SSB protein
  • Species:
  • Size:
    100 μg
  • Environmentally Preferable:
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    SSB
  • Protein/Peptide Name:
    Single-Stranded DNA Binding Protein (SSB)
  • Purity:
    >95%
  • Molecular Weight:
    Consists of four identical 18.9 kDa subunits
  • Shipping Temperature:
    Gel pack
  • Cat. No.:
    75838-160
  • Supplier no.:
    9220-100

Specifications

About this item

Single-stranded DNA binding protein (SSB) binds with high affinity in a cooperative manner to single-stranded DNA and does not bind well to double-stranded DNA. After binding single-stranded DNA, SSB destabilizes helical duplexes, thereby allowing DNA polymerases to access their substrate more easily.