Anti-IgM Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
76040-194
- Antibody Type:Secondary
- Antigen Name:Immunoglobulin M mu
- Antigen Symbol:IgM
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Mouse
- Size:2 mg
- Epitope:µ chain
- Storage Temperature:Store at 2-8 °C
- Cat. No.:76040-194
Goat anti-Mouse IgM (mu chain) - Affinity Pure, FITC Conjugate
Type: Secondary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope: µ chain
Host: Goat
Isotype:
Reactivity: Mouse