Plant Advance™ PCR Kit, BioVision
Catalog # 76005-440
Supplier: Avantor
CAS Number:
Specifications
- Description:Plant Advance™ PCR Kit
- Size:25 preps
- Cat. No.:76005-440
Specifications
About this item
Plant Advance™ PCR Kit, for PCR in a premixed and optimized format that simplifies the Plant PCR workflow. Plant Advance™ PCR Kit offers the ultimate convenience for plant PCR
- No time-consuming DNA purification steps
Application: Gene/transgene detection