Order Entry
United States
ContactUsLinkComponent
Nick translation DNA labeling system 2.0, Enzo Life Sciences
Nick translation DNA labeling system 2.0, Enzo Life Sciences
Catalog # 76013-836
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Nick translation DNA labeling system 2.0, Enzo Life Sciences
Catalog # 76013-836
Supplier:  Enzo Life Sciences
Supplier Number:  GEN111-0050
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Host:
  • Description:
    Nick translation DNA Labeling System 2.0
  • Size:
    50 tests
  • Environmentally Preferable:
  • Cat. No.:
    76013-836

Specifications

About this item

Nick translation system for preparing FISH probes

  • Ready-to-use Nick Translation Enzyme Mix minimizes pipetting error
  • Short labeling time - only 1 hour
  • Validated with fluorophore, biotin, and digoxigenin labeled dUTPs

The Nick Translation DNA Labeling System 2.0 provides a simple and efficient method for generating labeled DNA

The Nick Translation DNA Labeling System 2.0 can accommodate a wide range of fluorophore-labeled, biotin-labeled, and digoxigenin-labeled nucleotides. In addition to choice of label, the kit design allows the user to optimize incorporation and product size by adjusting the ratio of labeled-dUTP to dTTP. The ready-to-use NT Enzyme Mix is user friendly and minimizes error from pipetting.

Probes labeled by nick translation can be used in many different hybridization techniques including: in situ hybridization (ISH), fluorescent in situ hybridization (FISH), screening gene banks by colony or plaque hybridization, DNA or RNA transfer hybridization, and reassociation kinetic studies.

Contains NT Enzyme Mix 250 µL, Reaction Buffer 250 µL, dNTP Mix (dATP, dGTP, dCTP) 250 µL, dTTP 180 µL, Stop Buffer 250 µL, Nuclease-free Water 5 mL.