MULTIVIEW® PLUS (mouse-HRP/rabbit-AP) IHC Kit (Brown/Green), Enzo Life Sciences
75993-474
:
- Size:150 tests
- Brand Name:Enzo Life Sciences
- Cat. No.:75993-474
Superior multiplex IHC kit for efficient detection of multiple antigens in a single tissue sample.
- Biotin-free nanopolymer-based system for superior multiplex antigen detection
- Complete kit includes HIGHDEF® Chromogens for sensitive, crisp 2-color development
- Preserves precious tissue while reducing reagent and labor costs
- Expandable with additional chromogens (sold separately)
The MULTIVIEW® PLUS (mouse-HRP/rabbit-AP) IHC Kit is a non-biotin one-step detection system suitable for detecting and localizing antigens in formalin-fixed paraffin-embedded tissues.
Contains Antigen Retrieval Reagent, pH 9 (10X), ISH/IHC Peroxidase Block, Antibody Blocker/Diluent, MULTIVIEW® PLUS HRP (Anti-Mouse) Reagent, MULTIVIEW® PLUS AP (Anti-Rabbit) Reagent, HIGHDEF® DAB Chromogen, HIGHDEF® DAB Substrate, HIGHDEF® Green AP Chromogen, HIGHDEF® Green AP Substrate, Hematoxylin, Empty Mixing Bottle.