About this item
Interleukin-21 receptor (IL-21R) is a type I transmembrane glycoprotein within the class I cytokine receptor family, type 4 subfamily. Complex formation between IL-21R and the common gamma chain (gammac) is required for signaling. IL-21R is expressed mainly on B cells (highest on mature, activated, follicular and germinal center B cells), NK cells and activated T cells, but is also found on dendritic cells, alternatively activated macrophages, intestinal lamina propria fibroblasts and epithelial cells and keratinocytes. B cell IL-21R engagement induces Blimp1 (which mediates plasma cell differentiation) and is important for memory responses. IL-21R engagement on mouse NK cells enhances their cytotoxic activity and IFN-gamma production. IL-21R engagement on CD8+ T cells aids control of viral infection and tumor growth; IL-21R is also necessary for sufficient numbers of regulatory T cells to combat chronic inflammation. IL-21R expression is often upregulated in allergic skin inflammation, systemic lupus erythematosus and diffuse large B cell lymphoma (DLBCL).
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Conjugation:Unconjugated
- Protein Function:Interleukins
- Protein/Peptide Type:Recombinant
- Source:CHO cells
- Species:Mouse
- Storage Conditions:–20 °C
- Endotoxin Content:<0.06EU/µg
- Biological Activity:Shows the biological function of the IL-21R moiety and exerts a prolonged circulating half-life caused by the modified Fc domain.
- Gene ID:AAL82636.1
- Protein Synonyms:CD360|Interleukin-21 Receptor|NILR
- Protein/Peptide Name:IL-21R
- Purity:98%
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.2µm-filtered solution in PBS.
- Shipping Temperature:-20 °C, Blue Ice