Order Entry
United States
ContactUsLinkComponent
Mouse Recombinant CD279 (from CHO cells)
Mouse Recombinant CD279 (from CHO cells)
Catalog # 102981-538
Supplier:  Adipogen
CAS Number:  
Mouse Recombinant CD279 (from CHO cells)
Catalog # 102981-538
Supplier:  Adipogen
Supplier Number:  CHI-MF-110PD1-C100
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Mouse
  • Size:
    100 µg
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    <0.06EU/µg
  • Gene ID:
    NP_032824.1
  • Protein Synonyms:
    PD-1|Programmed Cell Death Protein 1
  • Protein/Peptide Name:
    CD279
  • Purity:
    98%
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.2µm-filtered solution in PBS.
  • Shipping Temperature:
    -20 °C, Blue Ice
  • Cat. No.:
    102981-538
  • Supplier no.:
    MF110PD1C100

Specifications

About this item

CD279 (Programmed Cell Death Protein 1; PD-1) is a type I transmembrane protein belonging to the CD28/CTLA-4 family of immunoreceptors that mediate signals for regulating immune responses. Members of the CD28/CTLA-4 family have been shown to either promote T cell activation (CD28 and ICOS) or downregulate T cell activation (CTLA-4 and PD-1). CD279 is expressed on activated T cells, B cells, myeloid cells and on a subset of thymocytes. In vitro, ligation of CD279 inhibits TCR-mediated T cell proliferation and production of IL-1, IL-4, IL-10 and IFN-gamma. In addition, CD279 ligation also inhibits BCR mediated signaling. CD279 deficient mice have a defect in peripheral tolerance and spontaneously develop autoimmune diseases.