Order Entry
United States
ContactUsLinkComponent
Mouse Recombinant CD276 (from CHO cells)
Mouse Recombinant CD276 (from CHO cells)
Catalog # 102981-522
Supplier:  Adipogen
CAS Number:  
Mouse Recombinant CD276 (from CHO cells)
Catalog # 102981-522
Supplier:  Adipogen
Supplier Number:  CHI-MF-110B7H3-C100
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Mouse
  • Size:
    100 µg
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    <0.06EU/µg
  • Gene ID:
    NP_598744.1
  • Protein Synonyms:
    B7-H3
  • Protein/Peptide Name:
    CD276
  • Purity:
    98%
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.2µm-filtered solution in PBS.
  • Shipping Temperature:
    -20 °C, Blue Ice
  • Cat. No.:
    102981-522
  • Supplier no.:
    MF110B7H3C100

Specifications

About this item

CD276 (B7-H3) is a member of the B7/CD28 superfamily of costimulatory molecules serving as an accessory modulator of T cell response. B7 family molecules, which are expressed on antigen-presenting cells and display extracellular regions containing immunoglobulin (Ig) variable (V)- and constant (C)-like domains, are known to modulate T cell receptor (TCR)-mediated T cell activation by providing co-signals that are either stimulatory or inhibitory. B7-H3 provides a stimulatory signal to T cells. However, recent studies suggest a negative regulatory role for B7-H3 in T cell responses. B7-H3 inhibited T cell proliferation mediated by antibody to T cell receptor or allogeneic antigen-presenting cells. B7-H3 is a negative regulator that preferentially affects T(H)1 responses. B7-H3 may play an important role in muscle-immune interactions, providing further evidence of the active role of muscle cells in local immunoregulatory processes. Recently, B7-H3 expression has also been found in a variety of different human cancers, including prostate cancer, clear cell renal cell carcinoma (ccRCC), non-small-cell lung cancer (NSCLC), pancreatic cancer, gastric cancer, ovarian cancer, colorectal cancer (CRC) and urothelial cell carcinoma. B7-H3 was expressed in some human cancers and correlated with poor outcome of cancer patients.