Order Entry
United States
ContactUsLinkComponent
Human Recombinant CD137 (from E. coli)
Human Recombinant CD137 (from E. coli)
Catalog # 102981-486
Supplier:  Adipogen
CAS Number:  
Human Recombinant CD137 (from E. coli)
Catalog # 102981-486
Supplier:  Adipogen
Supplier Number:  CHI-HR-200CD137-C025
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    25 µg
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    <0.1EU/µg
  • Gene ID:
    AAH06196.1
  • Protein Synonyms:
    CD137 Antigen|Tumor Necrosis Factor Receptor Superfamily Member 9|T Cell Antigen ILA|TNFRSF9|MGC2172|ILA|CDw137|4-1BB|4-1BB Ligand Receptor T Cell|Antigen 4-1BB Homolog
  • Protein/Peptide Name:
    CD137
  • Purity:
    95%
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from a concentrated sterile solution containing 50mM Tris-HCl buffer (pH 8.0) and 500mM NaCl.
  • Shipping Temperature:
    -20 °C, Blue Ice
  • Cat. No.:
    102981-486
  • Supplier no.:
    HR200CD137C025

Specifications

About this item

Human CD137 (4-1BB) is a costimulatory molecule of the tumor necrosis factor (TNF) receptor superfamily. The glycoprotein 4-1BB is expressed mainly on activated CD4+ and CD8+ T cells and binds to a high-affinity ligand (4-1BBL) expressed on several antigen-presenting cells such as macrophages and activated B cells. Upon ligand binding, 4-1BB is associated with the tumor receptor-associated factors (TRAF), the adaptor protein and mediates downstream signaling events including the activation of NF-kappaB and cytokine production. 4-1BB signaling either by binding to 4-1BBL or by antibody ligation delivers signals for T cell activation and growth as well as monocyte proliferation and B cell survival, and plays a important role in the amplification of T cell-mediated immune responses. In contrast, it can also enhance activation-induced T cell apoptosis when triggered by engagement of the TCR/CD3 complex. In addition, the 4-1BB/4-1BBL costimulatory pathway has been shown to augment secondary CTL responses to several viruses and increase antitumor immunity. 4-1BB is therefore a promising candidate for immunotherapy of human cancer.