Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:CHO cells
- Species:Mouse
- Size:50 µg
- Storage Conditions:–20 °C
- Endotoxin Content:<0.06EU/µg
- Gene ID:NP_033986
- Protein/Peptide Name:CD83
- Purity:98%
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.2µm-filtered solution in PBS.
- Shipping Temperature:-20 °C, Blue Ice
- Cat. No.:102981-474
- Supplier no.:MF110CD83C050
Mouse CD83 is a 30-35kDa member of the Siglec (or sialic-acid-binding immunoglobulin-like lectin) family of transmembrane proteins. CD83 is a primary marker for dendritic cells. It is also found on B cells, neutrophils, monocytes and macrophages. Except for dendritic cells, CD83 expression is often transient. CD83 binds to sialic acids on target cells. Membrane CD83 promotes T cell proliferation, particularly of CD8+ cytotoxic T cells. Soluble CD83 is immunosuppressive and blocks T cell activation. On monocytes, CD83 is suggested to drive monocytes into a fibrocyte phenotype. A lack of membrane-expressed CD83 leads to an unusual IL-4/ IL-10 producing CD4+ T cell phenotype.