Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:CHO cells
- Species:Mouse
- Size:100 µg
- Storage Conditions:–20 °C
- Endotoxin Content:<0.06EU/µg
- Gene ID:NP_033985.3
- Protein Synonyms:B7-1
- Protein/Peptide Name:CD80
- Purity:98%
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.2µm-filtered solution in PBS.
- Shipping Temperature:-20 °C, Blue Ice
- Cat. No.:102981-472
- Supplier no.:MF110CD80C100
CD80 (B7-1) and CD86 (B7-2) together with their receptors CD28 and CTLA-4 constitute one of the dominant costimulatory pathways that regulate T cell and B cell responses. Although both CTLA-4 and CD28 can bind to the same ligands, CTLA-4 binds to B7-1 and B7-2 with a 20-100 fold higher affinity than CD28 and is involved in the down-regulation of the immune response. B7-1 is expressed on activated B cells, activated T cells, and macrophages. B7-2 is constitutively expressed on interdigitating dendritic cells, Langerhans cells, peripheral blood dendritic cells, memory B cells and germinal center B cells. Additionally, B7-2 is expressed at low levels on monocytes and can be up-regulated through interferon-gamma. B7-1 and B7-2 are both members of the immunoglobulin superfamily. It has been observed that both human and mouse B7-1 and B7-2 can bind to either human or mouse CD28 and CTLA-4, suggesting that there are conserved amino acids which form the B7-1/B7-2/CD28/CTLA-4 critical binding sites.