Order Entry
United States
ContactUsLinkComponent
Human Recombinant NF-kappaB (active) (from Insect Cells (Sf21))
Human Recombinant NF-kappaB (active) (from Insect Cells (Sf21))
Catalog # 102980-378
Supplier:  Adipogen
CAS Number:  
Human Recombinant NF-kappaB (active) (from Insect Cells (Sf21))
Catalog # 102980-378
Supplier:  Adipogen
Supplier Number:  AG-40T-0021-C002
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    Insect cells (Sf21)
  • Species:
    Human
  • Size:
    2 µg
  • Storage Conditions:
    –80 °C
  • Biological Activity:
    ~1ng is required for high mobility shift assay.~5ng are required for reconstituted transcription assays.
  • Gene ID:
    P19838
  • Protein Synonyms:
    Nuclear Factor NF-kappaB p50 Subunit
  • Protein/Peptide Name:
    NF-kappaB (active)
  • Purity:
    95%
  • Concentration:
    Lot dependent (approx. 0.2mg/ml)
  • Formulation:
    Liquid. In 50mM TRIS-HCl, pH 7.5, containing 100mM sodium chloride, 0.2% NP-40, 50-100mM imidazole and 10% glycerol.
  • Shipping Temperature:
    -80 °C, Dry Ice
  • Cat. No.:
    102980-378
  • Supplier no.:
    AG40T0021C002

Specifications

About this item

NF-kappaB is a pleiotropic transcription factor present in almost all cell types and is the endpoint of a series of signal transduction events that are initiated by a vast array of stimuli related to many biological processes such as inflammation, immunity, differentiation, cell growth, tumorigenesis and apoptosis. NF-kappaB is a homo- or heterodimeric complex formed by the Rel-like domain-containing proteins RELA/p65, RELB, NFKB1/p105, NFKB1/p50, REL and NFKB2/p52. The heterodimeric p65-p50 complex is the most abundant complex. The dimers bind at kappaB sites in the DNA of their target genes and the individual dimers have distinct preferences for different kappaB sites that they can bind with distinguishable affinity and specificity. Different dimer combinations act as transcriptional activators or repressors, respectively. NF-kappaB complexes are held in the cytoplasm in an inactive state complexed with members of the NF-kappaB inhibitor (I-kappaB) family. In a conventional activation pathway, I-kappaB is phosphorylated by I-kappaB kinases (IKKs) in response to different activators, subsequently degraded thus liberating the active NF-kappaB complex which translocates to the nucleus. NF-kappaB heterodimeric p65-p50 and RelB-p50 complexes are transcriptional activators. The NF-kappaB p50-p50 homodimer is a transcriptional repressor, but can act as a transcriptional activator when associated with BCL3.