About this item
Beclin-1 (Bcl-2 interacting coiled-coil protein; ATG6) is a multifunctional protein, well-known as a key regulator of autophagy and required for the initiation step in autophagosome formation. It interacts with several cofactors (Atg14L, UVRAG, Bif-1, rubicon, Ambra1, HMGB1, nPIST, VMP1, SLAM, IP3R, PINK and survivin) to regulate the lipid kinase PI3K protein and promote formation of PIK3C3/VPS34-containing complexes. These interactions induce autophagy, which is a process of programmed cell survival increased during periods of cell stress and extinguished during the cell cycle. Furthermore, beclin-1 is implicated in numerous biological processes, including adaptation to stress, development, endocytosis, cytokinesis, immunity, infection, tumorigenesis, aging and cell death. Beclin-1 is tightly regulated by diverse mechanisms, such as epigenetic silencing, microRNA regulation, post-translational modifications and protein-protein interactions. Multiple diseases are associated with deficiency or malfunction of beclin-1, including cancer and neurodegeneration.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:–20 °C
- Gene ID:NCBI link NM_003766.3
- Protein Synonyms:ATG6|BECN1|GT197
- Protein/Peptide Name:Beclin-1
- Purity:85%
- Molecular Weight:~54kDa (SDS-PAGE)
- Concentration:0.1mg/ml after reconstitution.
- Formulation:Lyophilized from 25mM TRIS-HCl, pH 8.0, containing 100mM glycine.
- Shipping Temperature:-20 °C, Blue Ice