Order Entry
United States
ContactUsLinkComponent
Human Recombinant Frizzled-5 (from HEK293 cells)
Human Recombinant Frizzled-5 (from HEK293 cells)
Supplier:  Adipogen
Certificates
Human Recombinant Frizzled-5 (from HEK293 cells)
Supplier:  Adipogen

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    <0.01EU/µg
  • Biological Activity:
    Inhibits Wnt3a activation in a stoechiometric manner.
  • Gene ID:
    Q13467
  • Protein Synonyms:
    Fz-5|FzE5|hFz5
  • Protein/Peptide Name:
    Frizzled-5
  • Purity:
    95%
  • Molecular Weight:
    ~50kDa (SDS-PAGE)
  • Endotoxin Level:
    Low
  • Concentration:
    1mg/ml after reconstitution.
  • Formulation:
    Lyophilized. Contains PBS.
  • Shipping Temperature:
    -20 °C, Blue Ice

Specifications

Frequently Bought Together