About this item
Interleukin 6 (IL-6) is a pleiotropic alpha-helical cytokine that plays important roles in acute phase reactions, inflammation, hematopoiesis, bone metabolism and cancer progression. IL-6 activity is essential for the transition from acute inflammation to either acquired immunity or chronic inflammatory disease. It is secreted by multiple cell types as a 22kDa-28kDa phosphorylated and variably glycosylated molecule. IL-6 induces signaling through a cell surface heterodimeric receptor complex composed of a ligand binding subunit (IL-6R) and a signal transducing subunit (gp130). IL-6 is a key factor for the growth of plasma cells.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Conjugation:Unconjugated
- Protein Function:Interleukins
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Storage Conditions:–20 °C
- Endotoxin Content:<0.01EU/µg
- Biological Activity:Promotes growth of hybridoma/plasma cells at concentration <0.1ng /ml.
- Gene ID:P08505
- Protein Synonyms:Interleukin-6|B Cell Hybridoma Growth Factor|Interleukin HP-1
- Protein/Peptide Name:IL-6
- Purity:95%
- Molecular Weight:50kDa
- Endotoxin Level:Low
- Concentration:0.1 mg /ml after reconstitution
- Formulation:Lyophilized. Contains PBS.
- Shipping Temperature:-20 °C, Blue Ice