About this item
Progranulin (PGRN) is a widely expressed pluripotent growth factor which plays a role in processes such as development, wound repair and inflammation by activating signaling cascades that control cell cycle progression and cell motility. Its function in the central nervous system is of interest, as mutations in the PGRN gene were found in cases of frontotemporal degeneration (FTLD). In addition, PGRN has also been linked to tumorigenesis. Progranulin is a biomarker for FTLD, other types of Alzheimer‘s Disease (AD) and potentially for MCI (Mild Cognitive Impairment). Additionally, PGRN is described as a new ligand of TNF receptors and a potential therapeutic against inflammatory disease like arthritis.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:<5EU/mg
- Biological Activity:Activates ERK phosphorylation in THP-1 and MCF10A cells and induces cell migration of THP-1 monocytic leukemia cells.
- Gene ID:P28799
- Protein Synonyms:PC Cell-derived Growth Factor|Proepithelin|PEPI
- Protein/Peptide Name:Progranulin
- Purity:98%
- Molecular Weight:~76kDa (SDS-PAGE)
- Endotoxin Level:Low
- Concentration:After reconstitution:for 10µg size: 0.1mg/mlfor 50µg size: 1 mg/ml
- Formulation:Lyophilized. Contains PBS.
- Shipping Temperature:-20 °C, Blue Ice