Human Recombinant IFN-gamma
Catalog # 76003-386
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Protein/Peptide Type:Recombinant
- Species:Human
- Size:100 µg
- Protein Synonyms:Interferon-γ
- Protein/Peptide Name:IFN-gamma
- Purity:>98% (SDS-PAGE gel & HPLC)
- Molecular Weight:~16.8 kDa
- Formulation:Lyophilized from 10mM sodium phosphate, pH 7.4.
- Cat. No.:76003-386
Specifications
About this item
Produced in E.coli.