Order Entry
United States
Orders LinkContactUsLinkComponent
Nodularin, linear

Nodularin, linear

100 µg
Catalog # 76002-000
Supplier:   Enzo Life Sciences
Supplier #:  350-182-C100
Nodularin, linear
Catalog # 76002-000
Supplier:  Enzo Life Sciences
CAS Number:  118399-22-7
Nodularin, linear
Nodularin, linear

100 µg

CAS Number:  118399-22-7
Price
$325.91
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Precursor of the highly toxic cyclic pentapeptide, nodularin.

Isolated from Nodularia spumigena.

Isolated from Nodularia spumigena.

Linear nodularin has been demonstrated to be a biosynthetic precursor of the highly toxic cyclic pentapeptide, nodularin. It is an intermediate of biodegradation by natural populations of bacteria in aquatic samples.

Linear nodularin has been demonstrated to be a biosynthetic precursor of the highly toxic cyclic pentapeptide, nodularin. It is an intermediate of biodegradation by natural populations of bacteria in aquatic samples.

  • CAS Number:118399-22-7
  • Formula:C₄₁H₆₀N₈O₁₀
  • Molecular Weight:824.977 g/mol
Specifications

Specifications

  • Size:
    100 µg

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.