Rat Recombinant NGAL
Catalog # 76003-332
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Protein/Peptide Type:Recombinant
- Species:Rat
- Size:1 ml
- Protein Synonyms:Neutrophil gelatinase-associated lipocalin|Siderocalin|Lipocalin 2|Oncogene 24p3
- Protein/Peptide Name:NGAL
- Concentration:400pg/ml
- Formulation:Liquid. In 0.01 M phosphate buffer, pH 7.4, containing 0.14 M NaCl, 0.5% BSA, 0.5% Tween 20, 0.0016% Phenol red, and 0.1% Proclin 950.
- Cat. No.:76003-332
Specifications
About this item
Produced in E. coli.