Order Entry
United States
ContactUsLinkComponent
Recombinant Dengue NS1 ST3
Recombinant Dengue NS1 ST3
Catalog # 76003-314
Supplier:  Enzo Life Sciences
CAS Number:  
Recombinant Dengue NS1 ST3
Catalog # 76003-314
Supplier:  Enzo Life Sciences
Supplier Number:  PRT106-0100
CAS Number:  

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
  • Species:
  • Size:
    100 µg
  • Environmentally Preferable:
  • Protein Synonyms:
    non-structural protein|viral|strain Sri Lanka D3/H/IMTSSA-SRI/2000/1266|DENV1-NS1
  • Protein/Peptide Name:
    Dengue NS1 ST3
  • Purity:
    >95% (SDS-PAGE)
  • Formulation:
    Liquid. In DPBS pH 7.4.
  • Purification:
    Purified protein.
  • Cat. No.:
    76003-314

Specifications

About this item

Dengue virus NS1 protein serotype 3 presented in its native folded state with all post-translational modifications to deliver optimal antigenicity.

Produced in HEK 293 cells. Dengue Viral NS1 Serotype 3 (DENV1-NS1) from strain Sri Lanka D3/H/IMTSSA-SRI/2000/1266.