Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Symbol:DT
- Clonality:Monoclonal
- Clone:1C3
- Conjugation:Unconjugated
- Isotype:IgG2a kappa
- Reactivity:Multiple,Human
- Size:20 µg
- Form:Solid
- Shipping Temperature:-20° C
- Immunogen:3-(p-hydroxyphenyl) propionic acid dimer-conjugated keyhole limpet hemocyanin.
- Cat. No.:102979-290
- Supplier no.:JAIMDT020P
Specifications
About this item
Tyrosine is one of the major targets of protein oxidation. Dityrosine (DT) is known to be formed when tyrosine is damaged by free radicals, such as reactive oxygen species (ROS), metal-catalyzed oxidation, ultraviolet irradiation, and peroxidases. Dityrosine has been found in atherosclerotic lesions, and lipofuscin of pyramidal neurons of aged human brains. Dityrosine is a specific biomarker for protein oxidation. Dityrosine is reported to exist in various tissues and was also recently detected in urine samples. It is expected to be a novel protein oxdation marker, which is non-invasively detectable.
Type: Primary
Antigen: DT
Clonality: Monoclonal
Clone: 1C3
Conjugation: Unconjugated
Epitope:
Host:
Isotype: Mouse IgG2akappa
Reactivity: All, Human