To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Symbol:CD81
- Clonality:Monoclonal
- Clone:1.3.3.22
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Form:liquid
- Storage Temperature:+4°C
- Shipping Temperature:-20° C
- Immunogen:Human B cell line from Burkitt lymphoma.
- Cat. No.:102512-866
- Supplier no.:ANC302020
Human CD81 is a member of the tetraspan family of leukocyte cell surface molecules which are involved in signal transduction, cell-cell adhesion and cellular activation. Association of CD81 with integrins appears to be functionally important in the regulation of cell adhesion and activation. Recent work with CD81-null mice indicates CD81 is important for development of humoral immune response.
Type: Primary
Antigen: CD81
Clonality: Monoclonal
Clone: 1.3.3.22
Conjugation: Unconjugated
Epitope:
Host:
Isotype: Mouse IgG1
Reactivity: Human