Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Control
- Clonality:Monoclonal
- Clone:MOPC31C
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Isotype:IgG1 kappa
- Western Blot:Yes
- Size:100 µg
- Form:liquid
- Storage Temperature:+4°C
- Shipping Temperature:-20° C
- Cat. No.:102512-774
- Supplier no.:ANC278010
Immunoglobulin expressed by the mouse plasmacytoma MOPC 31C is an IgG1kappa isotype of unknown specificity. It is used as a negative control when working with human cells.
Type: Control
Antigen:
Clonality: Monoclonal
Clone: MOPC31C
Conjugation: Unconjugated
Epitope:
Host:
Isotype: Mouse IgG1kappa
Reactivity: