Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Symbol:CD64
- Clonality:Monoclonal
- Clone:10.1
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Isotype:IgG1
- Reactivity:Primates,Human,Dog
- Western Blot:Yes
- Size:100 µg
- Form:liquid
- Storage Temperature:+4°C
- Shipping Temperature:-20° C
- Immunogen:Human rheumatoid synovial fluid cells/monocytes.
- Cat. No.:102512-522
- Supplier no.:ANC216020
Specifications
About this item
Human CD64 is a high affinity receptor for monomeric human IgG1 and IgG3 which is expressed on macrophages, monocytes and gamma-interferon induced neutrophils. CD64 plays an important role in clearance of immune complexes and in antibody dependent cytoxicity.
Type: Primary
Antigen: CD64
Clonality: Monoclonal
Clone: 10.1
Conjugation: Unconjugated
Epitope:
Host:
Isotype: Mouse IgG1
Reactivity: Dog, Human, Primate