To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:CD59
- Clonality:Monoclonal
- Clone:BRA-10G
- Conjugation:Biotin
- Flow Cytometry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:liquid
- Storage Temperature:+4°C
- Shipping Temperature:-20° C
- Immunogen:Human K562 tumor cells.
- Cat. No.:102512-498
- Supplier no.:ANC211030
Specifications
About this item
Human CD59 is a GPI anchored molecule that protects cells from complement-mediated lysis. Studies are progressing to determine the regulatory mechanism for CD59 expression. A CD59-like protein has been discovered on the causative agent of Amebic encephalitis.
Type: Primary
Antigen: CD59
Clonality: Monoclonal
Clone: BRA-10G
Conjugation: Biotin
Epitope:
Host:
Isotype: Mouse IgG1kappa
Reactivity: Human