To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Symbol:CD57
- Clonality:Monoclonal
- Clone:NK-1
- Flow Cytometry:Yes
- Isotype:IgM
- Reactivity:Human
- Size:120 tests
- Form:liquid
- Storage Temperature:+4°C
- Shipping Temperature:-20° C
- Immunogen:Human PBL’s.
- Cat. No.:102512-482
- Supplier no.:ANC209040
Human CD57 originally called HNK-1, is a glycoprotein found on 15-20 % of PBL’s (peripheral blood lymphocytes) including 60 % of NK cells and a subset of T cells. The immune regulation role of CD57 positive PBL’s expressing high levels of CD8 is being investigated.
Type: Primary
Antigen: CD57
Clonality: Monoclonal
Clone: NK-1
Conjugation: FITC
Epitope:
Host:
Isotype: Mouse IgM
Reactivity: Human