To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:CD54
- Clonality:Monoclonal
- Clone:15.2
- ELISA:Yes
- Flow Cytometry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:liquid
- Storage Temperature:+4°C
- Shipping Temperature:-20° C
- Immunogen:Human monocytes.
- Cat. No.:102512-442
- Supplier no.:ANC205820
Specifications
About this item
Human CD54 (ICAM-1) mediates cell adhesion by binding to the integrins CD11a/CD18 (LFA-1) and to CD11b/CD18 (Mac-1). CD54 expression on resting peripheral blood leukocytes is weak but is upregulated on activated T and B lymphocytes and on monocytes.
- preservative free
Type: Primary
Antigen: CD54
Clonality: Monoclonal
Clone: 15.2
Conjugation: Preservative Free
Epitope:
Host:
Isotype: Mouse IgG1kappa
Reactivity: Human